Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diamino-5-bromopyrimidin-4-ol ≥96%

Supplier: MATRIX SCIENTIFIC MS

077017-500MG 077017-1G 077017-5G
102664-196EA 115.38 USD InStock
102664-196 102664-198 102664-200
2,6-Diamino-5-bromopyrimidin-4-ol ≥96%
2,6-Diamino-5-bromopyrimidin-4-ol
Hazard: Irritant


Formula: C₄H₅BrN₄O
MW: 205.02 g/mol
Melting Pt: 256…257 °C
MDL Number: MFCD00023262
CAS Number: 6312-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty