Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-2H,4H,5H,6H,7H-pyrazolo[4,3-b]pyridine-4-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

077055-500MG 077055-1G
102664-342EA 529.23 USD InStock
102664-342 102664-344
tert-Butyl-2H,4H,5H,6H,7H-pyrazolo[4,3-b]pyridine-4-carboxylate ≥95%
tert-Butyl-2H,4H,5H,6H,7H-pyrazolo[4,3-b]pyridine-4-carboxylate
Hazard: Irritant


Formula: C₁₁H₁₇N₃O₂
Melting Pt: 101…103 °C
MDL Number: MFCD22383945
CAS Number: 1393845-78-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty