Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(4-Methylphenyl)amino]-4-oxobutanoic acid

Supplier: MATRIX SCIENTIFIC MS

077115-500MG 077115-1G 077115-5G
102664-524EA 161.54 USD InStock
102664-524 102664-526 102664-528
4-[(4-Methylphenyl)amino]-4-oxobutanoic acid
4-[(4-Methylphenyl)amino]-4-oxobutanoic acid
Hazard: Irritant


Formula: C₁₁H₁₃NO₃
MDL Number: MFCD00205701
CAS Number: 37600-44-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty