Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(4-Nitrophenyl)amino]-4-oxobutanoic acid

Supplier: MATRIX SCIENTIFIC MS

077136-500MG 077136-1G 077136-5G
102664-648EA 161.54 USD InStock
102664-648 102664-650 102664-652
4-[(4-Nitrophenyl)amino]-4-oxobutanoic acid
4-[(4-Nitrophenyl)amino]-4-oxobutanoic acid
Hazard: Irritant


Formula: C₁₀H₁₀N₂O₅
MDL Number: MFCD00031339
CAS Number: 5502-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty