Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate

Supplier: MATRIX SCIENTIFIC MS

077385-500MG 077385-1G 077385-5G
102665-948EA 258.46 USD InStock
102665-948 102665-950 102665-952
Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate
Ethyl-5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carboxylate
Hazard: Irritant


Formula: C₁₂H₁₂ClN₃O₂
MW: 265.7 g/mol
MDL Number: MFCD00973905
CAS Number: 14678-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty