Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine-8-carboxamide

Supplier: MATRIX SCIENTIFIC MS

077578-500MG 077578-1G 077578-5G
102666-964EA 516.92 USD InStock
102666-964 102666-966 102666-968
Imidazo[1,2-a]pyridine-8-carboxamide
Imidazo[1,2-a]pyridine-8-carboxamide
Hazard: Irritant


Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD17215835
CAS Number: 144675-82-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty