Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tricyclo[4.3.1.1~3,8~]undecan-4-one

Supplier: MATRIX SCIENTIFIC MS

077622-500MG 077622-1G 077622-5G
102667-160EA 615.38 USD InStock
102667-160 102667-162 102667-164
Tricyclo[4.3.1.1~3,8~]undecan-4-one
Tricyclo[4.3.1.1~3,8~]undecan-4-one
Hazard: Irritant


Formula: C₁₁H₁₆O
MDL Number: MFCD11100000
CAS Number: 24669-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty