Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Carboxymethyl)-4,5-dimethoxybenzoic acid

Supplier: MATRIX SCIENTIFIC MS

077647-500MG 077647-1G 077647-5G
102667-298EA 516.92 USD InStock
102667-298 102667-300 102667-302
2-(Carboxymethyl)-4,5-dimethoxybenzoic acid
2-(Carboxymethyl)-4,5-dimethoxybenzoic acid
Hazard: Irritant


Formula: C₁₁H₁₂O₆
MDL Number: MFCD03447205
CAS Number: 3809-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty