Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[(1-Methyl-1H-pyrrol-2-yl)methyl]cyclopropanamine

Supplier: MATRIX SCIENTIFIC MS

080325-500MG 080325-1G 080325-5G
102669-106EA 553.85 USD InStock
102669-106 102669-108 102669-110
N-[(1-Methyl-1H-pyrrol-2-yl)methyl]cyclopropanamine
N-[(1-Methyl-1H-pyrrol-2-yl)methyl]cyclopropanamine
Hazard: Irritant


Formula: C₉H₁₄N₂
MW: 150.23 g/mol
MDL Number: MFCD07409353
CAS Number: 713501-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty