Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(4-methylphenyl)-3-oxopropanoate

Supplier: MATRIX SCIENTIFIC MS

080529-500MG 080529-1G 080529-5G
102669-990EA 95.38 USD InStock
102669-990 102669-992 102669-994
Methyl-3-(4-methylphenyl)-3-oxopropanoate
Methyl-3-(4-methylphenyl)-3-oxopropanoate
Hazard: Irritant


Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
MDL Number: MFCD03424778
CAS Number: 22027-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty