Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(3-chlorobenzyl)-3-oxobutanoate

Supplier: MATRIX SCIENTIFIC MS

080537-500MG 080537-1G 080537-5G
102670-036EA 129.23 USD InStock
102670-036 102670-038 102670-040
Ethyl-2-(3-chlorobenzyl)-3-oxobutanoate
Ethyl-2-(3-chlorobenzyl)-3-oxobutanoate
Hazard: Irritant


Formula: C₁₃H₁₅ClO₃
MDL Number: MFCD16329451
CAS Number: 30292-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty