Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-5-(3-nitrophenyl)thiophene-2-carboxylate

Supplier: MATRIX SCIENTIFIC MS

080642-500MG 080642-1G 080642-5G
102670-534EA 258.46 USD InStock
102670-534 102670-536 102670-538
Ethyl-3-amino-5-(3-nitrophenyl)thiophene-2-carboxylate
Ethyl-3-amino-5-(3-nitrophenyl)thiophene-2-carboxylate
Hazard: Irritant


Formula: C₁₃H₁₂N₂O₄S
MDL Number: MFCD04621479
CAS Number: 730949-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty