Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Bromopropyl)-1H-imidazole hydrobromide

Supplier: MATRIX SCIENTIFIC MS

080946-500MG 080946-1G
102672-032EA 438.46 USD InStock
102672-032 102672-034
1-(3-Bromopropyl)-1H-imidazole hydrobromide
1-(3-Bromopropyl)-1H-imidazole hydrobromide
Hazard: Irritant


Formula: C₆H₁₀Br₂N₂
MDL Number: MFCD18917307
CAS Number: 144385-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty