Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-bromo-2-oxobutanoate

Supplier: MATRIX SCIENTIFIC MS

080992-500MG 080992-1G 080992-5G
102672-270EA 149.23 USD InStock
102672-270 102672-272 102672-274
Ethyl-3-bromo-2-oxobutanoate
Ethyl-3-bromo-2-oxobutanoate
Hazard: Irritant


Formula: C₆H₉BrO₃
MDL Number: MFCD19440735
CAS Number: 57332-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty