Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6,7-dimethyl-4-hydroxypteridine ≥97%

Supplier: MATRIX SCIENTIFIC MS

081459-1G 081459-10G
102834-988EA 200 USD InStock
102834-988 102834-990
2-Amino-6,7-dimethyl-4-hydroxypteridine ≥97%
2-Amino-6,7-dimethyl-4-hydroxypteridine
Hazard: Irritant


Formula: C₈H₉N₅O
CAS Number: 611-55-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty