Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-9-ethyl carbazole (AEC) ≥97%

Supplier: MATRIX SCIENTIFIC MS

081494-10G 081494-50G
102835-072EA 90.77 USD InStock
102835-072 102835-074
3-Amino-9-ethyl carbazole (AEC) ≥97%
3-Amino-9-ethyl carbazole (AEC)
Hazard: Irritant


Formula: C₁₄H₁₄N₂
MW: 210.28
MDL Number: MFCD00004964
CAS Number: 132-32-1
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty