Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Furaldehyde ≥97%

Supplier: MATRIX SCIENTIFIC MS

081667-1G 081667-10G 081667-50G
102835-516EA 84.62 USD InStock
102835-516 102835-518 102835-520
3-Furaldehyde ≥97%
3-Furaldehyde
Hazard: Irritant


Formula: C₅H₄O₂
MW: 96.09 g/mol
MDL Number: MFCD00010424
CAS Number: 498-60-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty