Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexanemethylamine ≥97%

Supplier: MATRIX SCIENTIFIC MS

081966-1G 081966-10G 081966-50G
102836-224EA 72.31 USD InStock
102836-224 102836-226 102836-228
Cyclohexanemethylamine ≥97%
Cyclohexanemethylamine
Hazard: Flammable/Corrosive


Formula: C₇H₁₅N
MDL Number: MFCD00001520
CAS Number: 3218-02-8
UN: 2735
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty