Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-3,5-dinitrobenzoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

082029-1G 082029-10G 082029-50G
102836-366EA 56.92 USD InStock
102836-366 102836-368 102836-370
4-Methyl-3,5-dinitrobenzoic acid ≥97%
4-Methyl-3,5-dinitrobenzoic acid
Hazard: Irritant


Formula: C₈H₆N₂O₆
MW: 226.14 g/mol
MDL Number: MFCD00007175
CAS Number: 16533-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty