Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Tris(acetyloxy)benzoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

082224-1G 082224-10G
102836-854EA 569.23 USD InStock
102836-854 102836-856
3,4,5-Tris(acetyloxy)benzoic acid ≥97%
3,4,5-Tris(acetyloxy)benzoic acid
Hazard: Irritant


Formula: C₁₃H₁₂O₈
Melting Pt: 163…165 °C
MDL Number: MFCD00017226
CAS Number: 6635-24-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty