Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Ethoxybenzoyl)propionic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

082255-1G 082255-10G
102836-932EA 127.69 USD InStock
102836-932 102836-934
3-(4-Ethoxybenzoyl)propionic acid ≥97%
3-(4-Ethoxybenzoyl)propionic acid
Hazard: Irritant


Formula: C₁₂H₁₄O₄
MW: 222.24
MDL Number: MFCD00100659
CAS Number: 53623-37-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty