Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzylthio)nicotinic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

082492-1G 082492-10G 082492-50G
102837-454EA 72.31 USD InStock
102837-454 102837-456 102837-458
2-(Benzylthio)nicotinic acid ≥98%
2-(Benzylthio)nicotinic acid
Hazard: Irritant


Formula: C₁₃H₁₁NO₂S
MW: 245.30 g/mol
MDL Number: MFCD00661380
CAS Number: 112811-90-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty