Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromopyruvic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

083008-1G 083008-10G 083008-50G
102838-632EA 21.54 USD InStock
102838-632 102838-634 102838-636
Bromopyruvic acid ≥97%
Bromopyruvic acid
TCA listed: T
Hazard: Corrosive/Refrigerate


Formula: BrH₂CC(O)COOH
MW: 166.96 g/mol
Boiling Pt: 249
Melting Pt: 75…80 °C
MDL Number: MFCD00002587
CAS Number: 1113-59-3
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty