Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Allyloxypropionic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

083054-1G 083054-10G
102838-796EA 129.23 USD InStock
102838-796 102838-798
3-Allyloxypropionic acid ≥97%
3-Allyloxypropionic acid
Hazard: Irritant


Formula: C₆H₁₀O₃
MDL Number: MFCD00039537
CAS Number: 22577-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty