Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichlorophenyl isocyanate ≥97%

Supplier: MATRIX SCIENTIFIC MS

084305-1G 084305-10G
102842-034EA 101.54 USD InStock
102842-034 102842-036
2,3-Dichlorophenyl isocyanate ≥97%
2,3-Dichlorophenyl isocyanate
Hazard: Irritant


Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 247
MDL Number: MFCD00002000
CAS Number: 41195-90-8
UN: 2250
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty