Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxybenzonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS

084460-1G 084460-10G 084460-50G
102842-462EA 56.92 USD InStock
102842-462 102842-464 102842-466
3-Methoxybenzonitrile ≥98%
3-Methoxybenzonitrile
Hazard: Irritant


Formula: C₈H₇NO
MDL Number: MFCD00001801
CAS Number: 1527-89-5
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty