Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chloro-2-methylphenoxy)propanoic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

084769-1G 084769-10G
102843-324EA 353.85 USD InStock
102843-324 102843-326
2-(4-Chloro-2-methylphenoxy)propanoic acid ≥97%
2-(4-Chloro-2-methylphenoxy)propanoic acid
Hazard: Irritant


Formula: C₁₀H₁₁ClO₃
MW: 214.65 g/mol
MDL Number: MFCD00002648
CAS Number: 93-65-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty