Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate ≥97%

Supplier: MATRIX SCIENTIFIC MS

084913-10G 084913-50G
102843-718EA 116.92 USD InStock
102843-718 102843-720
N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate ≥97%
N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate
Hazard: Irritant


Formula: C₁₁H₁₈N₂O
MW: 292.35 g/mol
CAS Number: 25646-77-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty