Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Butoxyacetanilide ≥97%

Supplier: MATRIX SCIENTIFIC MS

084972-1G 084972-10G 084972-50G
102843-876EA 101.54 USD InStock
102843-876 102843-878 102843-880
4'-Butoxyacetanilide ≥97%
4'-Butoxyacetanilide
Hazard: Irritant


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD00027197
CAS Number: 23563-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty