Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-[(cyanomethyl)thio]benzoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083355-1G 083355-10G
102839-544EA 113.85 USD InStock
102839-544 102839-546
Methyl-2-[(cyanomethyl)thio]benzoate ≥97%
Methyl-2-[(cyanomethyl)thio]benzoate
Hazard: Irritant


Formula: C₁₀H₉NO₂S
MDL Number: MFCD00068076
CAS Number: 57601-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty