Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-bromo-2-chlorobenzoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083373-1G 083373-10G 083373-50G
102839-604EA 15.38 USD InStock
102839-604 102839-606 102839-608
Ethyl-5-bromo-2-chlorobenzoate ≥97%
Ethyl-5-bromo-2-chlorobenzoate
Hazard: Irritant


Formula: C₉H₈BrClO₂
Boiling Pt: 156-7°C
Density: 1.48
Flash Pt: 110°C
MDL Number: MFCD00013635
CAS Number: 76008-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty