Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-benzyl-4-piperidinecarboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083557-5G 083557-25G 083557-100G
102840-082EA 46.15 USD InStock
102840-082 102840-084 102840-086
Ethyl-1-benzyl-4-piperidinecarboxylate ≥97%
Ethyl-1-benzyl-4-piperidinecarboxylate
Hazard: Irritant


Formula: C₁₅H₂₁NO₂
MW: 247.34 g/mol
MDL Number: MFCD00040748
CAS Number: 24228-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty