Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl cyclopropanecarboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083620-1G 083620-10G 083620-50G
102840-250EA 15.38 USD InStock
102840-250 102840-252 102840-254
Ethyl cyclopropanecarboxylate ≥97%
Ethyl cyclopropanecarboxylate
Hazard: Flammable


Formula: C₆H₁₀O₂
MW: 114.15 g/mol
Boiling Pt: 133…134 °C
Density: 0.96
Flash Pt: 18 °C
MDL Number: MFCD00001282
CAS Number: 4606-07-9
UN: 3272
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty