Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl phenylcyanoacetate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083688-10G 083688-50G
102840-418EA 169.23 USD InStock
102840-418 102840-420
Ethyl phenylcyanoacetate ≥97%
Ethyl phenylcyanoacetate
Hazard: Irritant


Formula: C₁₁H₁₁NO₂
Boiling Pt: 163-165°C/10 Torr
Melting Pt: 115-116°C
Density: 1.085/17°C
MDL Number: MFCD00001866
CAS Number: 4553-07-5
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty