Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-chloro-2-methylbutyrate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083713-1G 083713-10G
102840-508EA 86.15 USD InStock
102840-508 102840-510
Methyl-4-chloro-2-methylbutyrate ≥97%
Methyl-4-chloro-2-methylbutyrate
Hazard: Irritant


Formula: C₆H₁₁ClO₂
MW: 150.60 g/mol
MDL Number: MFCD00198001
CAS Number: 13888-03-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty