Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methylthiophene ≥97%

Supplier: MATRIX SCIENTIFIC MS

083898-1G 083898-10G 083898-50G
102841-014EA 72.31 USD InStock
102841-014 102841-016 102841-018
2-Chloro-5-methylthiophene ≥97%
2-Chloro-5-methylthiophene
Hazard: Irritant


Formula: C₅H₅ClS
MDL Number: MFCD01632145
CAS Number: 17249-82-0
UN: 1992
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty