Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-(2-thienyl)-1,3-thiazole ≥90%

Supplier: MATRIX SCIENTIFIC MS

085338-1G 085338-10G
102844-784EA 298.46 USD InStock
102844-784 102844-786
2-Methyl-4-(2-thienyl)-1,3-thiazole ≥90%
2-Methyl-4-(2-thienyl)-1,3-thiazole
Hazard: Irritant


Formula: C₈H₇NS₂
MDL Number: MFCD01116539
CAS Number: 21036-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty