Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Mercaptohexyl acetate ≥95%

Supplier: MATRIX SCIENTIFIC MS

086035-5G 086035-25G
102846-712EA 158.46 USD InStock
102846-712 102846-714
3-Mercaptohexyl acetate ≥95%
3-Mercaptohexyl acetate
Hazard: Irritant-Harmful/Moisture Sensitive/Stench


Formula: C₈H₁₆O₂S
MDL Number: MFCD00792516
CAS Number: 136954-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty