Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,7-Trimethyloctane-3,5-dione ≥97%

Supplier: MATRIX SCIENTIFIC MS

086042-1G 086042-10G
102846-740EA 92.31 USD InStock
102846-740 102846-742
2,2,7-Trimethyloctane-3,5-dione ≥97%
2,2,7-Trimethyloctane-3,5-dione
Hazard: Irritant


Formula: C₁₁H₂₀O₂
Boiling Pt: 44 °C (0.2 mmHg)
MDL Number: MFCD00511279
CAS Number: 69725-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty