Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propionamide ≥97%

Supplier: MATRIX SCIENTIFIC MS

086126-1G 086126-10G 086126-50G
102847-030EA 207.69 USD InStock
102847-030 102847-032 102847-034
Propionamide ≥97%
Propionamide
Hazard: Irritant


Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 213
Melting Pt: 76…78 °C
MDL Number: MFCD00008039
CAS Number: 79-05-0
Merck Index: 12,08009
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty