Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',3,3-Trimethylbutyrophenone ≥97%

Supplier: MATRIX SCIENTIFIC MS

086739-1G 086739-2G 086739-5G
102850-336EA 672.31 USD InStock
102850-336 102850-338 102850-340
3',3,3-Trimethylbutyrophenone ≥97%
3',3,3-Trimethylbutyrophenone
Hazard: Irritant


Formula: C₁₃H₁₈O
MDL Number: MFCD03841075
CAS Number: 898785-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty