Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexyl-2,3-dimethylphenyl ketone ≥97%

Supplier: MATRIX SCIENTIFIC MS

086827-1G 086827-2G 086827-5G
102850-864EA 672.31 USD InStock
102850-864 102850-866 102850-868
Cyclohexyl-2,3-dimethylphenyl ketone ≥97%
Cyclohexyl-2,3-dimethylphenyl ketone
Hazard: Irritant


Formula: C₁₅H₂₀O
MDL Number: MFCD03841355
CAS Number: 898769-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty