Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) ≥97%

Supplier: MATRIX SCIENTIFIC MS

086904-1G 086904-2G 086904-5G
102851-326EA 520 USD InStock
102851-326 102851-328 102851-330
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV) ≥97%
4'-Butyl-2,2,2-trifluoroacetophenone (carbonate ionophore IV)
Hazard: Refrigerate


Formula: C₁₂H₁₃F₃O
MW: 230.22 g/mol
MDL Number: MFCD00214157
CAS Number: 40739-44-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty