Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-7-oxoundecanoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

087090-1G 087090-2G 087090-5G
102852-436EA 592.31 USD InStock
102852-436 102852-438 102852-440
Ethyl-7-oxoundecanoate ≥97%
Ethyl-7-oxoundecanoate
Hazard: Irritant


Formula: C₁₃H₂₄O₃
MW: 228.33 g/mol
MDL Number: MFCD01311673
CAS Number: 227953-80-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty