Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-(2-oxo-2-phenylethyl)benzoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

087108-1G 087108-2G 087108-5G
102852-544EA 1044.62 USD InStock
102852-544 102852-546 102852-548
Ethyl-4-(2-oxo-2-phenylethyl)benzoate ≥97%
Ethyl-4-(2-oxo-2-phenylethyl)benzoate
Hazard: Irritant


Formula: C₁₇H₁₆O₃
MDL Number: MFCD07700162
CAS Number: 898776-62-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty