Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-5-carbohydrazide

Supplier: MATRIX SCIENTIFIC MS

112980-1G 112980-5G
102891-312EA 436.92 USD InStock
102891-312 102891-314
1H-Indazole-5-carbohydrazide
1H-Indazole-5-carbohydrazide
Hazard: Irritant


Formula: C₈H₈N₄O
MDL Number: MFCD11506092
CAS Number: 1005205-25-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty