Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-1-benzylazetidine-3,3-dicarboxylate

Supplier: MATRIX SCIENTIFIC MS

113391-1G 113391-5G
102892-044EA 461.54 USD InStock
102892-044 102892-046
Diethyl-1-benzylazetidine-3,3-dicarboxylate
Diethyl-1-benzylazetidine-3,3-dicarboxylate
Hazard: Irritant


Formula: C₁₆H₂₁NO₄
MW: 291.35 g/mol
MDL Number: MFCD17169917
CAS Number: 642411-11-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty