Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Dichloro-1H-indole

Supplier: MATRIX SCIENTIFIC MS

113393-1G 113393-5G
102892-052EA 930.77 USD InStock
102892-052 102892-054
5,6-Dichloro-1H-indole
5,6-Dichloro-1H-indole
Hazard: Irritant


Formula: C₈H₅Cl₂N
MDL Number: MFCD01075036
CAS Number: 121859-57-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty