Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Hydroxyethyl)hexamethylenimine ≥97%

Supplier: MATRIX SCIENTIFIC MS

113698-1G 113698-10G
102892-642EA 86.15 USD InStock
102892-642 102892-644
1-(2-Hydroxyethyl)hexamethylenimine ≥97%
1-(2-Hydroxyethyl)hexamethylenimine
Hazard: Irritant


Formula: C₈H₁₇NO
MDL Number: MFCD00020988
CAS Number: 20603-00-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty