Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylcyclopropanecarboxylic acid ≥97%

Supplier: MATRIX SCIENTIFIC MS

114549-1G 114549-10G
102894-274EA 86.15 USD InStock
102894-274 102894-276
2-Methylcyclopropanecarboxylic acid ≥97%
2-Methylcyclopropanecarboxylic acid
Hazard: Irritant


Formula: C₅H₈O₂
MDL Number: MFCD00001293
CAS Number: 29555-02-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty